Please login to your online account to display your discounted pricing

Mouse anti-VEGF R2/KDR/Flk-1, Clone: KDR657, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen VEGF R2/KDR/Flk-1
Clone KDR657
Concentration 1mg/mL
Immunogen Recombinant human VEGF-R2 protein
Gene Accession No. P35968
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23465701

 
novus biologicals canada
NBP23465701MG
0.1mg Each for $384.00

NBP23465702

 
novus biologicals canada
NBP23465702MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen VEGF R2/KDR/Flk-1
Clone KDR657
Concentration 1mg/mL
Immunogen Recombinant human VEGF-R2 protein
Gene Accession No. P35968
Gene ID (Entrez) 3791
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Functional Assay