Please login to your online account to display your discounted pricing

Mouse anti-VEGF R2/KDR/Flk-1, Clone: KDR657, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23465701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23465701

  • / Each
 

Description & Specifications

Specifications

Antigen VEGF R2/KDR/Flk-1
Gene Accession No. P35968
Host Species Mouse
Immunogen Recombinant human VEGF-R2 protein
Isotype IgG1, κ
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3791
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Applications ELISA
Applications Functional Assay
Purification Method Purified
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Reconstitution Protein A purified
Clone KDR657