missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-VEGF R2/KDR/Flk-1, Clone: 2C6, Novus Biologicals™

$109.00 - $309.05

Specifications

Antigen VEGF R2/KDR/Flk-1
Clone 2C6
Concentration 1.0mg/mL
Immunogen Partial recombinant human VEGFR2 protein (between amino acids 300-600) [UniProt P35968]
Gene Accession No. P35968
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23642901 Novus Biologicals Canada
NBP23642901MG
0.1mg Each for $309.05
NBP23642925 Novus Biologicals Canada
NBP2364290025MG
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

VEGF R2/KDR/Flk-1
1.0mg/mL
P35968
PBS. Protein G purified.
Primary
Murine
Unlabeled
Purified
2C6
Partial recombinant human VEGFR2 protein (between amino acids 300-600) [UniProt P35968]
3791
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
Flow (Cell Surface),Flow Cytometry (Cell Surface),Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Western Blot