missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-VEGF R1/Flt-1, Clone: FLT1/659, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen VEGF R1/Flt-1
Clone FLT1/659
Concentration 1mg/mL
Immunogen Recombinant human VEGF-R1 protein
Gene Accession No. P17948
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23461901 Novus Biologicals Canada
NBP23461901MG
0.1mg Each for $384.00
NBP23461902 Novus Biologicals Canada
NBP23461902MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

VEGF R1/Flt-1
1mg/mL
P17948
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
FLT1/659
Recombinant human VEGF-R1 protein
2321
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Functional Assay