Please login to your online account to display your discounted pricing

Mouse anti-VEGF R1/Flt-1, Clone: FLT1/658, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23461801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23461801

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Applications ELISA
Applications Functional Assay
Conjugate Unlabeled
Concentration 1mg/mL
Primary or Secondary Primary
Gene ID (Entrez) 2321
Clone FLT1/658
Immunogen Recombinant human VEGF-R1 protein
Cross Reactivity Human
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 0.1mg
Host Species Mouse
Isotype IgG1, κ
Monoclonal or Polyclonal Monoclonal
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Antigen VEGF R1/Flt-1
Gene Accession No. P17948
Reconstitution Protein A purified
Purification Method Purified