Please login to your online account to display your discounted pricing

Mouse anti-VE-Cadherin, Clone: 16B1, PE, Novus Biologicals™

$175.00 - $395.00

Specifications

Antigen VE-Cadherin
Clone 16B1
Immunogen The immunogen for this antibody is VE Cadherin.
Gene ID (Entrez) 1003
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14380801

 
novus biologicals canada
NBP14380801MG
0.1mg Each for $395.00

NBP14380803

 
novus biologicals canada
NBP1438080025MG
0.025mg Each for $175.00
Description & Specifications

Specifications

Antigen VE-Cadherin
Clone 16B1
Immunogen The immunogen for this antibody is VE Cadherin.
Gene ID (Entrez) 1003
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry