Please login to your online account to display your discounted pricing

Mouse anti-VCAM-1/CD106, Clone: MR106, Biotin, Novus Biologicals™

$296.00 - $912.00

Specifications

Antigen VCAM-1/CD106
Clone MR106
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD106.
Gene ID (Entrez) 7412
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14359701

 
novus biologicals canada
NBP14359701MG
0.1mg Each for $428.00

NBP14359705

 
novus biologicals canada
NBP143597005MG
0.05mg Each for $296.00

NBP1435975

 
novus biologicals canada
NBP14359705MG
0.5mg Each for $912.00
Description & Specifications

Specifications

Antigen VCAM-1/CD106
Clone MR106
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD106.
Gene ID (Entrez) 7412
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry