Please login to your online account to display your discounted pricing

Mouse anti-V beta 13.2 TCR, Clone: H132, PE, Novus Biologicals™

$175.00 - $362.00

Specifications

Antigen V beta 13.2 TCR
Clone H132
Immunogen The immunogen for this antibody is V beta 13.2 TCR.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20031625

 
novus biologicals canada
NBP20031625TESTS
25 Tests Each for $175.00

NBP200316

 
novus biologicals canada
NBP200316100TESTS
100 Tests Each for $362.00
Description & Specifications

Specifications

Antigen V beta 13.2 TCR
Clone H132
Immunogen The immunogen for this antibody is V beta 13.2 TCR.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry