Please login to your online account to display your discounted pricing

Mouse anti-UACA, Clone: AE-5, Novus Biologicals™

$139.00 - $461.00

Specifications

Antigen UACA
Clone AE-5
Concentration 0.2mg/mL
Immunogen Nuclei of myeloid leukemia biopsy cells.
Gene Accession No. Q9BZF9
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23644301

 
novus biologicals canada
NBP23644301MG
0.1mg Each for $354.00

NBP23644302

 
novus biologicals canada
NBP236443002MG
0.02mg Each for $139.00

NBP2364432

 
novus biologicals canada
NBP23644302MG
0.2mg Each for $461.00
Description & Specifications

Specifications

Antigen UACA
Clone AE-5
Concentration 0.2mg/mL
Immunogen Nuclei of myeloid leukemia biopsy cells.
Gene Accession No. Q9BZF9
Gene ID (Entrez) 55075
Formulation 10mM PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)