Please login to your online account to display your discounted pricing

Mouse anti-UACA, Clone: AE-5, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP236443002MG

 View all options for this product


For Research Use Only

Catalog No. NBP23644302


Description & Specifications


Antigen UACA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone AE-5
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation 10mM PBS with 0.05% BSA. Protein G purified.
Gene Accession No. Q9BZF9
Host Species Mouse
Immunogen Nuclei of myeloid leukemia biopsy cells.
Isotype IgG1
Purification Method Purified
Quantity 0.02mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 55075