Please login to your online account to display your discounted pricing

Mouse anti-UACA, Clone: AE-5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23644301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23644301

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Gene Accession No. Q9BZF9
Host Species Mouse
Immunogen Nuclei of myeloid leukemia biopsy cells.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 0.2mg/mL
Antigen UACA
Format Purified
Formulation 10mM PBS with 0.05% BSA. Protein G purified.
Isotype IgG1
Purification Method Purified
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55075
Clone AE-5
Conjugate Unlabeled