Please login to your online account to display your discounted pricing

Mouse anti-UACA, Clone: AE-5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23644302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2364432

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Mouse
Regulatory Status RUO
Format Purified
Gene ID (Entrez) 55075
Antigen UACA
Concentration 0.2mg/mL
Isotype IgG1
Conjugate Unlabeled
Formulation 10mM PBS with 0.05% BSA. Protein G purified.
Quantity 0.2mg
Immunogen Nuclei of myeloid leukemia biopsy cells.
Primary or Secondary Primary
Gene Accession No. Q9BZF9
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone AE-5
Purification Method Purified
Monoclonal or Polyclonal Monoclonal