Please login to your online account to display your discounted pricing

Mouse anti-TYRP1, Clone: TA99, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23472001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23472001

  • / Each
 

Description & Specifications

Specifications

Formulation 10mM PBS. Does not contain BSA. Azide Free.
Quantity 0.1mg
Host Species Mouse
Antigen TYRP1
Gene Accession No. P17643
Isotype IgG2a, κ
Purification Method Purified
Clone TA99
Cross Reactivity Human
Cross Reactivity Mouse
Concentration 1mg/mL
Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Immunogen SK-MEL-23 cells
Gene ID (Entrez) 7306