Please login to your online account to display your discounted pricing

Mouse anti-TYRP1, Clone: SPM456, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen TYRP1
Clone SPM456
Concentration 0.2mg/mL
Immunogen SK-MEL-23 cells
Gene Accession No. P17643
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23290701

 
novus biologicals canada
NBP23290701ML
0.1mL Each for $139.00

NBP23290710

 
novus biologicals canada
NBP23290710ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen TYRP1
Clone SPM456
Concentration 0.2mg/mL
Immunogen SK-MEL-23 cells
Gene Accession No. P17643
Gene ID (Entrez) 7306
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting