Please login to your online account to display your discounted pricing

Mouse anti-TYRP1, Clone: SPM456, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23290710ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23290710

  • / Each
 

Description & Specifications

Specifications

Antigen TYRP1
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM456
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P17643
Host Species Mouse
Immunogen SK-MEL-23 cells
Isotype IgG2a, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7306
Reconstitution Protein A purified