Please login to your online account to display your discounted pricing

Mouse anti-TYRP1, Clone: SPM456, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23290701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23290701

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen SK-MEL-23 cells
Regulatory Status RUO
Concentration 0.2mg/mL
Gene Accession No. P17643
Antigen TYRP1
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Format Purified
Purification Method Purified
Quantity 0.1mL
Gene ID (Entrez) 7306
Reconstitution Protein A purified
Clone SPM456
Isotype IgG2a, κ
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal