missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-TYRP1, Clone: SPM456, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen TYRP1
Clone SPM456
Concentration 1mg/mL
Immunogen SK-MEL-23 cells
Gene Accession No. P17643
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23477301 Novus Biologicals Canada
NBP23477301MG
0.1mg Each for $384.00
NBP23477302 Novus Biologicals Canada
NBP23477302MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

TYRP1
1mg/mL
P17643
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM456
SK-MEL-23 cells
7306
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2a, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot