Please login to your online account to display your discounted pricing

Mouse anti-TYRP1, Clone: SPM456, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23477302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23477302

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P17643
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Host Species Mouse
Immunogen SK-MEL-23 cells
Isotype IgG2a, κ
Quantity 0.2mg
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7306
Concentration 1mg/mL
Antigen TYRP1
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Primary or Secondary Primary
Test Specificity It reacts with a 75kDa melanocyte-specific gene product, identified as Tyrosinase-related protein-1 (TRP-1).
Clone SPM456