Please login to your online account to display your discounted pricing

Mouse anti-Tyrosinase, Clone: SPM360, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23290510ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23290510

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Antigen Tyrosinase
Format Purified
Formulation PBS with 0.05% BSA.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant tyrosinase protein
Quantity 1.0mL
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Gene Accession No. P14679
Isotype IgG2a, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 7299
Clone SPM360