Please login to your online account to display your discounted pricing

Mouse anti-Tyrosinase, Clone: SPM360, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23290501ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23290501

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P14679
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant tyrosinase protein
Isotype IgG2a, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Clone SPM360
Conjugate Unlabeled
Antigen Tyrosinase
Host Species Mouse
Quantity 0.1mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7299
Concentration 0.2mg/mL
Cross Reactivity Human