Please login to your online account to display your discounted pricing

Mouse anti-TTF1 / NKX2.1, Clone: SPM150, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23299910ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23299910

  • / Each
 

Description & Specifications

Specifications

Antigen TTF1 / NKX2.1
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Rat full length TTF-1 recombinant protein
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 7080
Reconstitution Protein A purified
Clone SPM150
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Gene Accession No. P43699
Monoclonal or Polyclonal Monoclonal
Test Specificity Shows broad species reactivity.
Concentration 0.2mg/mL