Please login to your online account to display your discounted pricing

Mouse anti-TTF1 / NKX2.1, Clone: SPM150, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23474202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23474202

  • / Each
 

Description & Specifications

Specifications

Antigen TTF1 / NKX2.1
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone SPM150
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P43699
Host Species Murine
Immunogen Rat full length TTF-1 recombinant protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7080
Test Specificity Recognizes a protein of 40kDa, identified as Thyroid transcription factor-1 (TTF-1). Expected to show broad species reactivity.
Reconstitution Protein A purified
Target Species Human
Target Species Murine
Target Species Rat