Please login to your online account to display your discounted pricing

Mouse anti-TTF1 / NKX2.1, Clone: SPM150, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23474201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23474201

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Rat full length TTF-1 recombinant protein
Regulatory Status RUO
Primary or Secondary Primary
Antigen TTF1 / NKX2.1
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P43699
Host Species Mouse
Purification Method Purified
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7080
Test Specificity Recognizes a protein of 40kDa, identified as Thyroid transcription factor-1 (TTF-1). Expected to show broad species reactivity.
Reconstitution Protein A purified
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG1, κ
Clone SPM150
Concentration 1mg/mL