Please login to your online account to display your discounted pricing

Mouse anti-TTF1 / NKX2.1, Clone: 8G7G3/1, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen TTF1 / NKX2.1
Clone 8G7G3/1
Concentration 1mg/mL
Immunogen Rat full length TTF-1 recombinant protein
Gene Accession No. P43699
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23454401

 
novus biologicals canada
NBP23454401MG
0.1mg Each for $384.00

NBP23454402

 
novus biologicals canada
NBP23454402MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen TTF1 / NKX2.1
Clone 8G7G3/1
Concentration 1mg/mL
Immunogen Rat full length TTF-1 recombinant protein
Gene Accession No. P43699
Gene ID (Entrez) 7080
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot