Please login to your online account to display your discounted pricing

Mouse anti-TTF1 / NKX2.1, Clone: 8G7G3/1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23454401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23454401

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen TTF1 / NKX2.1
Primary or Secondary Primary
Gene ID (Entrez) 7080
Test Specificity Expected to show broad species reactivity.
Conjugate Unlabeled
Host Species Mouse
Regulatory Status RUO
Immunogen Rat full length TTF-1 recombinant protein
Monoclonal or Polyclonal Monoclonal
Quantity 0.1mg
Concentration 1mg/mL
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Isotype IgG1, κ
Gene Accession No. P43699
Purification Method Purified
Clone 8G7G3/1