Please login to your online account to display your discounted pricing

Mouse anti-TSLP, Clone: 15B11.3, Novus Biologicals™

$278.00 - $418.00

Specifications

Antigen TSLP
Clone 15B11.3
Immunogen The immunogen for this antibody was TSLP.
Gene ID (Entrez) 85480
Formulation PBS [pH 7.2]
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP1433115M

 
novus biologicals canada
NBP143311005MG
0.05mg Each for $278.00

NBP1433115

 
novus biologicals canada
NBP14331105MG
0.5mg Each for $418.00
Description & Specifications

Specifications

Antigen TSLP
Clone 15B11.3
Immunogen The immunogen for this antibody was TSLP.
Gene ID (Entrez) 85480
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications ELISA