Please login to your online account to display your discounted pricing

Mouse anti-TSLP, Clone: 15B11.3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143311005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1433115M

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Antigen TSLP
Format Affinity Purified
Formulation PBS [pH 7.2]
Immunogen The immunogen for this antibody was TSLP.
Isotype IgG1
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Clone 15B11.3
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 85480