Please login to your online account to display your discounted pricing

Mouse anti-TSLP, Clone: 15B11.3, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP14331105MG

 View all options for this product


For Research Use Only

Catalog No. NBP1433115


Description & Specifications


Antigen TSLP
Applications ELISA
Clone 15B11.3
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Mouse
Immunogen The immunogen for this antibody was TSLP.
Isotype IgG1
Purification Method Affinity Purified
Quantity 0.5mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 85480