Please login to your online account to display your discounted pricing

Mouse anti-TRAIL/TNFSF10, Clone: RIK-2, Biotin, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen TRAIL/TNFSF10
Clone RIK-2
Immunogen Human TRAIL-transfected mouse cell line
Gene ID (Entrez) 8743
Formulation Phosphate buffer pH 7.2. Immunogen affinity purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14347401

 
novus biologicals canada
NBP14347401MG
0.1mg Each for $252.00

NBP14347403

 
novus biologicals canada
NBP1434740025MG
0.025mg Each for $131.00
Description & Specifications

Specifications

Antigen TRAIL/TNFSF10
Clone RIK-2
Immunogen Human TRAIL-transfected mouse cell line
Gene ID (Entrez) 8743
Formulation Phosphate buffer pH 7.2. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry