Please login to your online account to display your discounted pricing

Mouse anti-TRAIL R1/TNFRSF10A, Clone: DJR1, PE, Novus Biologicals™

$171.00 - $354.00

Specifications

Antigen TRAIL R1/TNFRSF10A
Clone DJR1
Immunogen Extracellular domain of DR4-human IgG1 Fc fusion protein
Gene ID (Entrez) 8797
Formulation Immunogen affinity purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200206

 
novus biologicals canada
NBP200206100TESTS
100 Tests Each for $354.00

NBP20020625

 
novus biologicals canada
NBP20020625TESTS
25 Tests Each for $171.00
Description & Specifications

Specifications

Antigen TRAIL R1/TNFRSF10A
Clone DJR1
Immunogen Extracellular domain of DR4-human IgG1 Fc fusion protein
Gene ID (Entrez) 8797
Formulation Immunogen affinity purified
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry