Please login to your online account to display your discounted pricing

Mouse anti-TRAIL R1/TNFRSF10A, Clone: DJR1, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP20020625TESTS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20020625

  • / Each
 

Description & Specifications

Specifications

Antigen TRAIL R1/TNFRSF10A
Formulation Immunogen affinity purified
Host Species Mouse
Applications Flow Cytometry
Immunogen Extracellular domain of DR4-human IgG1 Fc fusion protein
Isotype IgG1, κ
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 8797
Test Specificity DR4 (DJR1)
Clone DJR1
Conjugate PE
Cross Reactivity Human
Format Affinity Purified
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.