Please login to your online account to display your discounted pricing

Mouse anti-TRAIL R1/TNFRSF10A, Clone: DJR1, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200206100TESTS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP200206

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications Flow Cytometry
Immunogen Extracellular domain of DR4-human IgG1 Fc fusion protein
Quantity 100 Tests
Test Specificity DR4 (DJR1)
Cross Reactivity Human
Format Affinity Purified
Formulation Immunogen affinity purified
Isotype IgG1, κ
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 8797
Clone DJR1
Conjugate PE
Antigen TRAIL R1/TNFRSF10A
Purification Method Affinity Purified