Please login to your online account to display your discounted pricing

Mouse anti-TRAIL R1/TNFRSF10A, Clone: DJR1, Biotin, Novus Biologicals™

$171.00 - $321.00

Specifications

Antigen TRAIL R1/TNFRSF10A
Clone DJR1
Concentration 0.5mg/mL
Immunogen Extracellular domain of DR4-human IgG1 Fc fusion protein
Gene ID (Entrez) 8797
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14349403

 
novus biologicals canada
NBP1434940025MG
0.025mg Each for $171.00

NBP14349401

 
novus biologicals canada
NBP14349401MG
0.1mg Each for $321.00
Description & Specifications

Specifications

Antigen TRAIL R1/TNFRSF10A
Clone DJR1
Concentration 0.5mg/mL
Immunogen Extracellular domain of DR4-human IgG1 Fc fusion protein
Gene ID (Entrez) 8797
Formulation Phosphate buffer pH 7.2. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry