Please login to your online account to display your discounted pricing

Mouse anti-TRAIL R1/TNFRSF10A, Clone: DJR1, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1434940025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14349403

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Antigen TRAIL R1/TNFRSF10A
Format Affinity Purified
Formulation Phosphate buffer pH 7.2. Immunogen affinity purified.
Host Species Mouse
Applications Flow Cytometry
Immunogen Extracellular domain of DR4-human IgG1 Fc fusion protein
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 8797
Clone DJR1
Cross Reactivity Human
Monoclonal or Polyclonal Monoclonal
Test Specificity DR4 (DJR1)
Concentration 0.5mg/mL
Conjugate Biotin