Please login to your online account to display your discounted pricing

Mouse anti-TRAIL R1/TNFRSF10A, Clone: DJR1, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14349401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14349401

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Host Species Mouse
Storage Requirements Store at 4°C. Do not freeze.
Antigen TRAIL R1/TNFRSF10A
Concentration 0.5mg/mL
Conjugate Biotin
Immunogen Extracellular domain of DR4-human IgG1 Fc fusion protein
Quantity 0.1mg
Primary or Secondary Primary
Formulation Phosphate buffer pH 7.2. Immunogen affinity purified.
Gene ID (Entrez) 8797
Isotype IgG1, κ
Format Affinity Purified
Purification Method Affinity Purified
Cross Reactivity Human
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Test Specificity DR4 (DJR1)
Clone DJR1