Please login to your online account to display your discounted pricing

Mouse anti-TRA-1-60 (R), Clone: MG38, FITC, Novus Biologicals™

$192.00 - $354.00

Specifications

Antigen TRA-1-60 (R)
Clone MG38
Immunogen The immunogen for this antibody was Fluorescein CD205.
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14396603

 
novus biologicals canada
NBP1439660025MG
0.025mg Each for $192.00

NBP14396601

 
novus biologicals canada
NBP14396601MG
0.1mg Each for $354.00
Description & Specifications

Specifications

Antigen TRA-1-60 (R)
Clone MG38
Immunogen The immunogen for this antibody was Fluorescein CD205.
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry