Please login to your online account to display your discounted pricing

Mouse anti-TRA-1-60 (R), Clone: MG38, FITC, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP1439660025MG

 View all options for this product


For Research Use Only

Catalog No. NBP14396603


Description & Specifications


Antigen TRA-1-60 (R)
Applications Flow Cytometry
Clone MG38
Conjugate FITC
Cross Reactivity Human
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Mouse
Immunogen The immunogen for this antibody was Fluorescein CD205.
Isotype IgG2b, κ
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal