Please login to your online account to display your discounted pricing

Mouse anti-TRA-1-60 (R), Clone: MG38, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14396601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14396601

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Immunogen The immunogen for this antibody was Fluorescein CD205.
Isotype IgG2b, κ
Monoclonal or Polyclonal Monoclonal
Quantity 0.1mg
Cross Reactivity Human
Applications Flow Cytometry
Storage Requirements Store at 4°C. Do not freeze.
Formulation Phosphate buffer pH 7.2
Regulatory Status RUO
Primary or Secondary Primary
Antigen TRA-1-60 (R)
Purification Method Affinity Purified
Conjugate FITC
Format Affinity Purified
Clone MG38