Please login to your online account to display your discounted pricing

Mouse anti-TNIP1, Clone: 5C4, Novus Biologicals™

$285.00 - $406.00

Specifications

Antigen TNIP1
Clone 5C4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Naf1.
Gene ID (Entrez) 10318
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14313701

 
novus biologicals canada
NBP14313701MG
0.1mg Each for $406.00

NBP14313703

 
novus biologicals canada
NBP1431370025MG
0.025mg Each for $285.00
Description & Specifications

Specifications

Antigen TNIP1
Clone 5C4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Naf1.
Gene ID (Entrez) 10318
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting