Please login to your online account to display your discounted pricing

Mouse anti-TNIP1, Clone: 5C4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1431370025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14313703

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Formulation Aqueous buffer (contains stabilizer if necessary)
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Clone 5C4
Antigen TNIP1
Host Species Mouse
Immunogen The immunogen for this antibody was Naf1.
Quantity 0.025mg
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Isotype IgG1
Purification Method Affinity Purified
Gene ID (Entrez) 10318