Please login to your online account to display your discounted pricing

Mouse anti-TNIP1, Clone: 5C4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14313701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14313701

  • / Each
 

Description & Specifications

Specifications

Antigen TNIP1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone 5C4
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation Aqueous buffer (contains stabilizer if necessary)
Host Species Murine
Immunogen The immunogen for this antibody was Naf1.
Isotype IgG1
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 10318
Target Species Human