Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: TNF706 + P/T2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23430405ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23430405

  • / Each
 

Description & Specifications

Specifications

Quantity 0.5mL
Immunogen Recombinant human TNF-alpha (TNF706); A hexadecapeptide corresponding to aa115-130 of human TNF-alpha
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Reconstitution Protein A purified
Cross Reactivity Canine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Zebrafish
Purification Method Purified
Isotype IgG1, κ
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Primary or Secondary Primary
Concentration 0.2mg/mL
Antigen TNF-alpha
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Clone TNF706 + P/T2
Gene Accession No. P01375
Host Species Mouse
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 7124