Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: TNF706 + P/T2, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23472901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23472901

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Canine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Zebrafish
Format Purified
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Quantity 0.1mg
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1, κ
Gene Accession No. P01375
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7124
Antigen TNF-alpha
Regulatory Status RUO
Immunogen Recombinant human TNF-alpha (TNF706); A hexadecapeptide corresponding to aa115-130 of human TNF-alpha
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Concentration 1mg/mL
Purification Method Purified
Reconstitution Protein A purified
Clone TNF706 + P/T2
Conjugate Unlabeled