Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: TNF706, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen TNF-alpha
Clone TNF706
Concentration 1mg/mL
Immunogen Recombinant human TNF-alpha
Gene Accession No. P01375
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23472801

 
novus biologicals canada
NBP23472801MG
0.1mg Each for $384.00

NBP23472802

 
novus biologicals canada
NBP23472802MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen TNF-alpha
Clone TNF706
Concentration 1mg/mL
Immunogen Recombinant human TNF-alpha
Gene Accession No. P01375
Gene ID (Entrez) 7124
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Canine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Zebrafish
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation