Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: TNF656, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen TNF-alpha
Clone TNF656
Concentration 0.2mg/mL
Immunogen Recombinant human TNF-alpha
Gene Accession No. P01375
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23430701

 
novus biologicals canada
NBP23430701ML
0.1mL Each for $139.00

NBP23430705

 
novus biologicals canada
NBP23430705ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen TNF-alpha
Clone TNF656
Concentration 0.2mg/mL
Immunogen Recombinant human TNF-alpha
Gene Accession No. P01375
Gene ID (Entrez) 7124
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA