Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: TNF656, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23430705ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23430705

  • / Each
 

Description & Specifications

Specifications

Antigen TNF-alpha
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Recombinant human TNF-alpha
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 7124
Clone TNF656
Conjugate Unlabeled
Format Purified
Gene Accession No. P01375
Applications ELISA
Isotype IgG1, κ
Purification Method Purified
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Concentration 0.2mg/mL
Cross Reactivity Human