Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: TNF656, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23430701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23430701

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Antigen TNF-alpha
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01375
Immunogen Recombinant human TNF-alpha
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7124
Reconstitution Protein A purified
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Clone TNF656
Concentration 0.2mg/mL