missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: TNF656, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen TNF-alpha
Clone TNF656
Concentration 1mg/mL
Immunogen Recombinant human TNF-alpha
Gene Accession No. P01375
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23472701 Novus Biologicals Canada
NBP23472701MG
0.1mg Each for $384.00
NBP23472702 Novus Biologicals Canada
NBP23472702MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

TNF-alpha
1mg/mL
P01375
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
TNF656
Recombinant human TNF-alpha
7124
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Functional Assay