Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: TNF656, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23472701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23472701

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Functional Assay
Antigen TNF-alpha
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Immunogen Recombinant human TNF-alpha
Isotype IgG1, κ
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 7124
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Gene Accession No. P01375
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone TNF656