Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: TNF655, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23430605ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23430605

  • / Each
 

Description & Specifications

Specifications

Antigen TNF-alpha
Applications ELISA
Clone TNF655
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01375
Host Species Mouse
Immunogen Recombinant human TNF-alpha
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7124
Reconstitution Protein A purified